Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005696786.3 | XP_005696843.1 | 5 - 6 | 823 | 96 | |
GenBank mRNA | JO433239.2 | - | 5 - 6 | 732 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |