Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005696786.3 | XP_005696843.1 | 9 - 10 | 1449 | 83 | |
GenBank mRNA | JO433239.2 | - | 1 - 2 | 106 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |