Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMERB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005697522.2 | XP_005697579.2 | 2 - 3 | 399 | 66 | |
GenBank mRNA | JO634954.2 | - | 4 - 5 | 1577 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |