Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDFIC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018047180.1 | XP_017902669.1 | 3 - 4 | 770 | 91 | |
XM_018047181.1 | XP_017902670.1 | 3 - 4 | 767 | 91 | ||
XM_018047182.1 | XP_017902671.1 | 3 - 4 | 360 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |