Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC29A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018042501.1 | XP_017897990.1 | 5 - 6 | 852 | 75 | |
XM_018042502.1 | XP_017897991.1 | 6 - 7 | 901 | 75 | ||
GenBank mRNA | KA315623.1 | - | 5 - 6 | 885 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons |