Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005690840.2 | XP_005690897.2 | 2 - 3 | 318 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |