Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102171003 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692929.3 | XP_005692986.1 | 3 - 4 | 304 | 68 | |
GenBank mRNA | JO319208.2 | - | 3 - 4 | 290 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |