Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005694730.3 | XP_005694787.1 | 2 - 3 | 1826 | 112 | |
GenBank mRNA | JT749361.1 | - | 3 - 4 | 567 | 112 | |
KT899958.1 | - | 2 - 3 | 613 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Capra hircus Probe spans exons |