Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018054965.1 | XP_017910454.1 | 4 - 5 | 501 | 63 | |
GenBank mRNA | JO439582.2 | - | 2 - 3 | 684 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |