Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018058563.1 | XP_017914052.1 | 2 - 3 | 1322 | 89 | |
XM_018058564.1 | XP_017914053.1 | 2 - 3 | 835 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Capra hircus Probe spans exons |