Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005683052.3 | XP_005683109.1 | 3 - 4 | 354 | 76 | |
GenBank mRNA | KA343687.1 | - | 3 - 4 | 1398 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons |