Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LSM11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018050106.1 | XP_017905595.1 | 3 - 4 | 719 | 81 | |
GenBank mRNA | JT648815.1 | - | 3 - 4 | 352 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons |