Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005685487.2 | XP_005685544.1 | 2 - 3 | 367 | 99 | |
XM_005685488.3 | XP_005685545.1 | 2 - 3 | 506 | 99 | ||
GenBank mRNA | JO403893.2 | - | 2 - 3 | 1125 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |