Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFP90 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692179.3 | XP_005692236.1 | 2 - 3 | 427 | 71 | |
XM_013971259.2 | XP_013826713.1 | 2 - 3 | 428 | 71 | ||
XM_013971260.2 | XP_013826714.1 | 2 - 3 | 428 | 71 | ||
XM_013971261.2 | XP_013826715.1 | 2 - 3 | 427 | 71 | ||
XM_018062191.1 | XP_017917680.1 | 3 - 4 | 373 | 71 | ||
GenBank mRNA | JO449256.2 | - | 2 - 3 | 405 | 71 | |
JO611018.2 | - | 2 - 3 | 462 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons |