Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018066638.1 | XP_017922127.1 | 2 | 673 | 59 | |
XM_018066639.1 | XP_017922128.1 | 2 | 698 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Capra hircus Both primers and probe map within a single exon |