Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDIT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005699173.3 | XP_005699230.1 | 2 - 3 | 404 | 55 | |
GenBank mRNA | JO560162.2 | - | 2 - 3 | 420 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |