Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAL3ST4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005697831.3 | XP_005697888.2 | 2 - 3 | 436 | 97 | |
GenBank mRNA | JO624627.2 | - | 2 - 3 | 435 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |