Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018044744.1 | XP_017900233.1 | 2 - 3 | 584 | 73 | |
GenBank mRNA | JO449435.2 | - | 1 - 2 | 169 | 73 | |
JO581425.1 | - | 4 - 5 | 881 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons |