Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMOD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018046977.1 | XP_017902466.1 | 1 - 2 | 428 | 105 | |
GenBank mRNA | JO375752.2 | - | 1 - 2 | 98 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |