Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3RF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018055296.1 | XP_017910785.1 | 8 - 9 | 3037 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |