Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005690991.3 | XP_005691048.1 | 3 - 4 | 814 | 67 | |
XM_005690992.3 | XP_005691049.1 | 3 - 4 | 679 | 67 | ||
XM_013970432.2 | XP_013825886.1 | 3 - 4 | 646 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |