Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM4SF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018064535.1 | XP_017920024.1 | 4 - 5 | 737 | 68 | |
GenBank mRNA | KA325405.1 | - | 1 - 2 | 133 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Capra hircus Probe spans exons |