Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARGFX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005709423.2 | XP_005709480.2 | 5 - 6 | 1028 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |