Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSBPL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018038204.1 | XP_017893693.1 | 4 - 5 | 881 | 67 | |
GenBank mRNA | JO427547.1 | - | 8 - 9 | 2157 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |