Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRNB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005693503.2 | XP_005693560.2 | 2 - 3 | 269 | 69 | |
XM_013972198.2 | XP_013827652.2 | 2 - 3 | 254 | 69 | ||
XM_013972199.2 | XP_013827653.1 | 2 - 3 | 254 | 69 | ||
XM_018064583.1 | XP_017920072.1 | 2 - 3 | 254 | 69 | ||
GenBank mRNA | JT747035.1 | - | 3 - 4 | 193 | 69 | |
KA311468.1 | - | 2 - 3 | 162 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |