Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FXYD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005699038.3 | XP_005699095.1 | 4 - 5 | 231 | 103 | |
XM_013975553.2 | XP_013831007.1 | 4 - 5 | 207 | 103 | ||
GenBank mRNA | JO458816.2 | - | 4 - 5 | 195 | 103 | |
JT668021.1 | - | 3 - 4 | 119 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |