Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERMN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005676116.3 | XP_005676173.2 | 1 - 2 | 569 | 93 | |
GenBank mRNA | JO634162.2 | - | 2 - 3 | 3220 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Capra hircus Probe spans exons |