Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLAMF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018046366.1 | XP_017901855.1 | 3 - 4 | 755 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |