Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MANEAL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005678678.3 | XP_005678735.3 | 3 - 4 | 809 | 82 | |
GenBank mRNA | JO638869.2 | - | 1 - 2 | 1749 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |