Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005693112.2 | XP_005693169.1 | 1 - 2 | 1814 | 114 | |
GenBank mRNA | JT703675.1 | - | 2 - 3 | 1732 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Capra hircus Probe spans exons |