Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMGB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018044330.1 | XP_017899819.1 | 2 - 3 | 259 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |