Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5H12orf45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018048200.1 | XP_017903689.1 | 2 - 3 | 253 | 108 | |
GenBank mRNA | JO415844.2 | - | 2 - 3 | 474 | 108 | |
KA323509.1 | - | 2 - 3 | 235 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |