Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYNPO2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018042200.1 | XP_017897689.1 | 3 - 4 | 975 | 62 | |
GenBank mRNA | JO344206.2 | - | 3 - 4 | 971 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Capra hircus Probe spans exons |