Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LENEP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677413.3 | XP_005677470.3 | 1 - 2 | 670 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |