Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005699905.3 | XP_005699962.2 | 1 - 2 | 178 | 97 | |
GenBank mRNA | JT754452.1 | - | 5 - 6 | 2768 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |