Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018040662.1 | XP_017896151.1 | 3 - 4 | 682 | 61 | |
XM_018040668.1 | XP_017896157.1 | 3 - 4 | 594 | 61 | ||
XM_018040671.1 | XP_017896160.1 | 3 - 4 | 682 | 61 | ||
GenBank mRNA | JO402301.2 | - | 3 - 4 | 1014 | 61 | |
JO435706.2 | - | 3 - 4 | 419 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |