Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBX21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018064958.1 | XP_017920447.1 | 2 - 3 | 864 | 68 | |
GenBank mRNA | JO580568.1 | - | 1 - 2 | 222 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |