Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP5PB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677897.3 | XP_005677954.1 | 2 - 3 | 175 | 74 | |
GenBank mRNA | JO437631.2 | - | 2 - 3 | 211 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |