Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLIPR1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005679728.2 | XP_005679785.2 | 2 - 3 | 530 | 155 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |