Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018055206.1 | XP_017910695.1 | 4 - 5 | 1044 | 68 | |
GenBank mRNA | JO415472.2 | - | 4 - 5 | 951 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |