Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRMS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018057532.1 | XP_017913021.1 | 3 - 4 | 887 | 78 | |
GenBank mRNA | KA339345.1 | - | 5 - 6 | 2856 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |