Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD79A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692572.3 | XP_005692629.1 | 2 - 3 | 605 | 91 | |
GenBank mRNA | JO341774.2 | - | 3 - 4 | 655 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |