Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUOX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018048094.1 | XP_017903583.1 | 3 - 4 | 618 | 64 | |
XM_018048095.1 | XP_017903584.1 | 3 - 4 | 604 | 64 | ||
XM_018048096.1 | XP_017903585.1 | 4 - 5 | 685 | 64 | ||
XM_018048097.1 | XP_017903586.1 | 3 - 4 | 319 | 64 | ||
GenBank mRNA | EU418447.2 | - | 3 - 4 | 285 | 64 | |
JO429150.2 | - | 3 - 4 | 327 | 64 | ||
JO502728.2 | - | 3 - 4 | 340 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |