Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC31A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018052620.1 | XP_017908109.1 | 2 - 3 | 190 | 82 | |
XM_018052621.1 | XP_017908110.1 | 2 - 3 | 221 | 82 | ||
GenBank mRNA | JO501894.2 | - | 2 - 3 | 173 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |