Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APH1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677684.3 | XP_005677741.1 | 6 - 7 | 1134 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |