Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TERB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005686226.2 | XP_005686283.2 | 2 - 3 | 167 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |