Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PXMP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005688471.3 | XP_005688528.3 | 2 - 3 | 373 | 65 | |
GenBank mRNA | JO437404.2 | - | 2 - 3 | 395 | 65 | |
KA327044.1 | - | 2 - 3 | 880 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |