Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNK16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018039311.1 | XP_017894800.1 | 3 - 4 | 510 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |