Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018047287.1 | XP_017902776.1 | 5 - 6 | 3647 | 76 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Capra hircus Probe spans exons |