Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C7H19orf53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018051216.1 | XP_017906705.1 | 2 - 3 | 213 | 98 | |
GenBank mRNA | JO410851.2 | - | 2 - 3 | 223 | 98 | |
KA350847.1 | - | 2 - 3 | 197 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Capra hircus Probe spans exons |